missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (+/-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine

Product Code. 11482070
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 185057-50-5
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD03001673
InChI Key FPXJNSKAXZNWMQ-UHFFFAOYSA-N
Synonym 3-amino-1-n-cbz-pyrrolidine, 1-cbz-3-aminopyrrolidine, 1-cbz-3-amino pyrrolidine, 1-benzyloxycarbonyl-3-aminopyrrolidine, 3-amino-pyrrolidine-1-carboxylic acid benzyl ester, 1-n-cbz-3-aminopyrrolidine, 3-amino-1-cbz-pyrrolidine, phenylmethyl 3-aminopyrrolidinecarboxylate, 1-pyrrolidinecarboxylic acid, 3-amino-, phenylmethyl ester, n-cbz-3-amino-pyrrolidine
PubChem CID 2756614
IUPAC Name benzyl 3-aminopyrrolidine-1-carboxylate
SMILES C1CN(CC1N)C(=O)OCC2=CC=CC=C2

Specifications

Refractive Index 1.5485
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 220.27
Chemical Name or Material (+/-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'