missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-1-phenylbutane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a chemical intermediate. 1-Methyl-3-phenylpropylamine is used in the synthesis of N-substituted derivatives of (1-methyl-3- phenylpropyl)amine.
Solubility
Slightly soluble in water. Soluble in chloroform and ethyl acetate.
Notes
Air Sensitive. Store away from air, oxidizing agents and carbon dioxide. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is employed as a chemical intermediate. 1-Methyl-3-phenylpropylamine is used in the synthesis of N-substituted derivatives of (1-methyl-3- phenylpropyl)amine.
Solubility
Slightly soluble in water. Soluble in chloroform and ethyl acetate.
Notes
Air Sensitive. Store away from air, oxidizing agents and carbon dioxide. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Specifications
Melting Point | -50°C |
Density | 0.928 |
Boiling Point | 220°C to 222°C |
Flash Point | 97°C (206°F) |
Odor | Amine-like |
Refractive Index | 1.514 |
Quantity | 500 g |
UN Number | UN2922 |
Beilstein | 2413110 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'