missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-1H-1,2,4-triazole-5-carboxylic acid hydrate, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Specifications
Melting Point | 178°C to 183°C |
Color | White |
Loss on Drying | 2% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Quantity | 25 g |
Synonym | 3-amino-1,2,4-triazole-5-carboxylic acid, 5-amino-4h-1,2,4-triazole-3-carboxylic acid, 5-amino-1h-1,2,4-triazole-3-carboxylic acid, 1h-1,2,4-triazole-3-carboxylic acid, 5-amino, 5-amino-1,2,4-triazole-3-carboxylic acid, s-triazole-3-carboxylic acid, 5-amino, 5-amino-2h-1,2,4-triazole-3-carboxylic acid, 5-amino-1h-1,2,4 triazole-3-carboxylic acid, 5-amino-2h-1,2,4 triazole-3-carboxylic acid, 1h-1,4-triazole-3-carboxylic acid, 5-amino |
Formula Weight | 128.09 |
Percent Purity | ≥97.5% (on dry substance) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'