missing translation for 'onlineSavingsMsg'
Learn More

3'-Amino-2',3'-dideoxyadenosine, 99%, Thermo Scientific™

Product Code. 15436719
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7403-25-0
Color White to Yellow
Molecular Formula C10H14N6O2
MDL Number MFCD08703999
Quantity 1 g
Synonym 3'-amino-2',3'-dideoxyadenosine, 2',3'-dideoxy-3'-aminoadenosine, adenosine, 3'-amino-2',3'-dideoxy, 2s,3s,5r-3-amino-5-6-amino-9h-purin-9-yl tetrahydrofuran-2-yl methanol, 2s,3s,5r-3-amino-5-6-aminopurin-9-yl oxolan-2-yl methanol, 2s,3s,5r-3-amino-5-6-aminopurin-9-yl tetrahydrofuran-2-yl methanol, 3'-nh-3'-dado, 3'-amino-2'-deoxy-d-adenosine?, 3?-amino-2?,3?-dideoxyadenosine, 3'-amino-2', 3'-dideoxyadenosine
InChI Key MVAJRASUDLEWKZ-RRKCRQDMSA-N
SMILES N[C@H]1C[C@@H](O[C@@H]1CO)N1C=NC2=C(N)N=CN=C12
IUPAC Name [(2S,3S,5R)-3-amino-5-(6-aminopurin-9-yl)oxolan-2-yl]methanol
Molecular Weight (g/mol) 250.26
PubChem CID 165600
Formula Weight 250.26
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3'-Amino-2',3'-dideoxyadenosine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.