missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methylacrylaldehyde, 97%, Thermo Scientific™

Product Code. 10501492
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 30989-81-2
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD01566866
InChI Key OEZAAXHZEMTBOV-DUXPYHPUSA-N
Synonym 3-amino-2-methylacrylaldehyde, 3-amino-2-methylacrolein, 2e-3-amino-2-methylprop-2-enal, amino-alpha-methylacrolein, e-3-amino-2-methylprop-2-enal, 3-amino-2-methyl acrylaldehyde, 2-propenal, 3-amino-2-methyl-, e
PubChem CID 637539
IUPAC Name (E)-3-amino-2-methylprop-2-enal
SMILES CC(=CN)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula NH2CH=C(CH3)CHO
Quantity 10 g
Formula Weight 85.11
Percent Purity 97%
Chemical Name or Material 3-Amino-2-methylacrylaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'