missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methylphenol, 98%, Thermo Scientific Chemicals

Product Code. 15422789
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53222-92-7
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007787
InChI Key FLROJJGKUKLCAE-UHFFFAOYSA-N
Synonym 3-amino-o-cresol, phenol, 3-amino-2-methyl, 2-amino-o-cresol, 2-methyl-3-aminophenol, 3-hydroxy-2-methylaniline, aminokresol, o-cresol, 3-amino, pubchem15039, 2-methyl-3-amino-phenol, 3-amino-2-methyl phenol
PubChem CID 104448
IUPAC Name 3-amino-2-methylphenol
SMILES CC1=C(C=CC=C1O)N

Specifications

Melting Point 129°C to 130°C
Quantity 25 g
Beilstein 2716070
Formula Weight 123.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Amino-2-methylphenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'