missing translation for 'onlineSavingsMsg'
Learn More
3-Amino-3-(4-fluorophenyl)propionic acid, 96%, Thermo Scientific™ Product Code.: 11379765

3-Amino-3-(4-fluorophenyl)propionic acid, 96%, Thermo Scientific™

Product Code. 11379765
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11379765

Brand: Thermo Scientific Alfa Aesar L19799.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 325-89-3
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.182
MDL Number MFCD00181811
InChI Key CPGFMWPQXUXQRX-UHFFFAOYSA-N
Synonym 3-amino-3-4-fluorophenyl propanoic acid, 3-amino-3-4-fluorophenyl propionic acid, 3-amino-3-4-fluoro-phenyl-propionic acid, 3-p-fluorophenyl-dl-beta-alanine, r-3-p-fluorophenyl-beta-alanine, 3-4-fluorophenyl-beta-alanine, dl-3-amino-3-4-fluoro-phenyl-propionic acid, pubchem13896, pubchem17300, acmc-20a8bwShow MoreShow Less
PubChem CID 579885
IUPAC Name 3-amino-3-(4-fluorophenyl)propanoic acid
SMILES C1=CC(=CC=C1C(CC(=O)O)N)F

Specifications

Melting Point 214°C (decomposition)
Beilstein 3258498
Quantity 250 mg
Formula Weight 183.18
Percent Purity 96%
Chemical Name or Material 3-Amino-3-(4-fluorophenyl)propionic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-3-(4-fluorophenyl)propionic acid, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.