missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-bromo-1-methyl-1H-pyrazole, 97+%, Thermo Scientific Chemicals

Product Code. 11457204
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146941-72-2
Molecular Formula C4H6BrN3
Molecular Weight (g/mol) 176.02
MDL Number MFCD00663065
InChI Key WYPUMGAZSVZUGJ-UHFFFAOYSA-N
PubChem CID 2735283
SMILES CN1C=C(Br)C(N)=N1

Specifications

Melting Point 96°C to 100°C
Quantity 5 g
Synonym 4-bromo-1-methyl-1h-pyrazol-3-amine, 3-amino-4-bromo-1-methyl-1h-pyrazole, 3-amino-4-bromo-1-methylpyrazole, 1h-pyrazol-3-amine, 4-bromo-1-methyl, 4-bromo-1-methylpyrazole-3-ylamine, acmc-1bzbh, 1h-pyrazol-3-amine,4-bromo-1-methyl
IUPAC Name 4-bromo-1-methylpyrazol-3-amine
Formula Weight 176.02
Percent Purity ≥97%
Chemical Name or Material 3-Amino-4-bromo-1-methyl-1H-pyrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'