missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-bromo-1H-indazole, 95%, Thermo Scientific™

Product Code. 10833391
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 914311-50-5
Molecular Formula C7H6BrN3
Molecular Weight (g/mol) 212.05
MDL Number MFCD09909580
InChI Key IXXXOIXGQFPLIH-UHFFFAOYSA-N
Synonym 3-amino-4-bromo-1h-indazole, 4-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 4-bromo, 3-amino-4-bromoindazole, pubchem20681, acmc-209rbi, 4-bromo-1h-indazole-3-amine, 4-bromo-2h-indazol-3-ylamine, 4-bromo-1h-indazole-3-ylamine
PubChem CID 11975123
IUPAC Name 4-bromo-1H-indazol-3-amine
SMILES NC1=NNC2=CC=CC(Br)=C12

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 148.0°C to 150.0°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 212.05
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-bromo-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-4-bromo-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'