Learn More
3-Amino-4-bromo-1H-indazole, 95%, Thermo Scientific™
£141.90 - £202.00
Chemical Identifiers
CAS | 914311-50-5 |
---|---|
Molecular Formula | C7H6BrN3 |
Molecular Weight (g/mol) | 212.05 |
MDL Number | MFCD09909580 |
InChI Key | IXXXOIXGQFPLIH-UHFFFAOYSA-N |
Synonym | 3-amino-4-bromo-1h-indazole, 4-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 4-bromo, 3-amino-4-bromoindazole, pubchem20681, acmc-209rbi, 4-bromo-1h-indazole-3-amine, 4-bromo-2h-indazol-3-ylamine, 4-bromo-1h-indazole-3-ylamine |
PubChem CID | 11975123 |
IUPAC Name | 4-bromo-1H-indazol-3-amine |
SMILES | NC1=NNC2=CC=CC(Br)=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10189969
|
Thermo Scientific Acros
435790010 |
1 g | |||||||
10833391
|
Thermo Scientific Acros
435790050 |
5 g |
£202.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
914311-50-5 | |
212.05 | |
IXXXOIXGQFPLIH-UHFFFAOYSA-N | |
11975123 | |
NC1=NNC2=CC=CC(Br)=C12 |
C7H6BrN3 | |
MFCD09909580 | |
3-amino-4-bromo-1h-indazole, 4-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 4-bromo, 3-amino-4-bromoindazole, pubchem20681, acmc-209rbi, 4-bromo-1h-indazole-3-amine, 4-bromo-2h-indazol-3-ylamine, 4-bromo-1h-indazole-3-ylamine | |
4-bromo-1H-indazol-3-amine |
Specifications
914311-50-5 | |
100.0 | |
Gray to White | |
95% | |
MFCD09909580 | |
IXXXOIXGQFPLIH-UHFFFAOYSA-N | |
4-bromo-1H-indazol-3-amine | |
11975123 | |
≥94.0% | |
3-Amino-4-bromo-1H-indazole |
94.0 | |
147°C to 152°C | |
Authentic | |
C7H6BrN3 | |
3-amino-4-bromo-1h-indazole, 4-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 4-bromo, 3-amino-4-bromoindazole, pubchem20681, acmc-209rbi, 4-bromo-1h-indazole-3-amine, 4-bromo-2h-indazol-3-ylamine, 4-bromo-1h-indazole-3-ylamine | |
NC1=NNC2=CC=CC(Br)=C12 | |
212.05 | |
212.05 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- 3-Amino-4-bromo-1H-indazole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only