missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-bromobenzoic acid, 97%, Thermo Scientific™

Product Code. 15467108
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2840-29-1
Melting Point 226°C to 230°C
Molecular Formula C7H6BrNO2
MDL Number MFCD00579106
Quantity 1 g
Synonym 3-amino-4-bromo-benzoic acid, benzoic acid, 3-amino-4-bromo, 3-amino-4-bromobenzoicacid, acmc-1cpid, 4-brom-3-aminobenzoesaure, ksc494k6h, 3-amino-4-bromobenzoic acid, 3-amino-4-bromo benzoic acid
InChI Key TZFJADFQEBASQU-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(=O)O)N)Br
IUPAC Name 3-amino-4-bromobenzoic acid
Molecular Weight (g/mol) 216.034
PubChem CID 3418752
Formula Weight 216.03
Percent Purity 97%
Chemical Name or Material 3-Amino-4-bromobenzoic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'