Learn More
3-Amino-4-chlorophenol, 97%, Thermo Scientific Chemicals
£50.80 - £543.00
Chemical Identifiers
CAS | 16026-77-0 |
---|---|
Molecular Formula | C6H6ClNO |
Molecular Weight (g/mol) | 143.57 |
MDL Number | MFCD03427127 |
InChI Key | FWUALUHYKLDYAN-UHFFFAOYSA-N |
PubChem CID | 5070078 |
IUPAC Name | 3-amino-4-chlorophenol |
SMILES | C1=CC(=C(C=C1O)N)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15402188
|
Thermo Scientific Alfa Aesar
H55826.03 |
1 g |
£50.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15412188
|
Thermo Scientific Alfa Aesar
H55826.06 |
5 g | |||||||
15422188
|
Thermo Scientific Alfa Aesar
H55826.14 |
25 g | |||||||
Description
3-Amino-4-chlorophenol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic chemical intermediate for synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Amino-4-chlorophenol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic chemical intermediate for synthesis.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
16026-77-0 | |
143.57 | |
FWUALUHYKLDYAN-UHFFFAOYSA-N | |
3-amino-4-chlorophenol |
C6H6ClNO | |
MFCD03427127 | |
5070078 | |
C1=CC(=C(C=C1O)N)Cl |
Specifications
16026-77-0 | |
Brown to Orange | |
MFCD03427127 | |
Slightly soluble in water. | |
C1=CC(=C(C=C1O)N)Cl | |
143.57 | |
143.57 | |
Powder |
151°C to 154°C | |
C6H6ClNO | |
4-chloro-3-aminophenol, acmc-1bo5g, 2-chloro-5-hydroxyaniline, phenol,3-amino-4-chloro, 3-azanyl-4-chloranyl-phenol, phenol, 3-amino-4-chloro, 3-amino-4-chlorophenol | |
FWUALUHYKLDYAN-UHFFFAOYSA-N | |
3-amino-4-chlorophenol | |
5070078 | |
97% | |
3-Amino-4-chlorophenol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only