Learn More
3-Amino-4-cyanopyridine, 97%, Thermo Scientific™
£44.52 - £100.00
Chemical Identifiers
CAS | 78790-79-1 |
---|---|
Molecular Formula | C6H5N3 |
Molecular Weight (g/mol) | 119.13 |
MDL Number | MFCD04035596 |
InChI Key | PEZNQSQPDQLHPN-UHFFFAOYSA-N |
Synonym | 3-amino-4-cyanopyridine, 3-aminoisonicotinonitrile, 3-amino-isonicotinonitrile, 4-pyridinecarbonitrile, 3-amino, pubchem9759, 3-amino4-cyanopyridine, acmc-209pfc, 3-amino-4-cyano-pyridine, 3-aminopyridine4-carbonitrile, ksc497q6tShow More |
PubChem CID | 10678219 |
IUPAC Name | 3-aminopyridine-4-carbonitrile |
SMILES | NC1=C(C=CN=C1)C#N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
78790-79-1 | |
119.13 | |
PEZNQSQPDQLHPN-UHFFFAOYSA-N | |
10678219 | |
NC1=C(C=CN=C1)C#N |
C6H5N3 | |
MFCD04035596 | |
3-amino-4-cyanopyridine, 3-aminoisonicotinonitrile, 3-amino-isonicotinonitrile, 4-pyridinecarbonitrile, 3-amino, pubchem9759, 3-amino4-cyanopyridine, acmc-209pfc, 3-amino-4-cyano-pyridine, 3-aminopyridine4-carbonitrile, ksc497q6tShow More | |
3-aminopyridine-4-carbonitrile |
Specifications
78790-79-1 | |
C6H5N3 | |
UN3439 | |
PEZNQSQPDQLHPN-UHFFFAOYSA-N | |
3-aminopyridine-4-carbonitrile | |
10678219 | |
97% |
135°C to 138°C | |
MFCD04035596 | |
3-amino-4-cyanopyridine, 3-aminoisonicotinonitrile, 3-amino-isonicotinonitrile, 4-pyridinecarbonitrile, 3-amino, pubchem9759, 3-amino4-cyanopyridine, acmc-209pfc, 3-amino-4-cyano-pyridine, 3-aminopyridine4-carbonitrile, ksc497q6t | |
NC1=C(C=CN=C1)C#N | |
119.13 | |
119.13 | |
3-Amino-4-cyanopyridine |
Safety and Handling
GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P330-P361-P363-P501c
H301+H311-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.