missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-fluorobenzoic acid, 97%, Thermo Scientific™

Product Code. 15467738
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2365-85-7
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00130055
InChI Key WFSPEVFSRUTRCN-UHFFFAOYSA-N
Synonym 3-amino-4-fluorobenzoicacid, benzoic acid, 3-amino-4-fluoro, 5-carboxy-2-fluoroaniline, 3-amino-4-fluoro-benzoic acid, pubchem4690, acmc-1cilf, ksc207o2p, benzoicacid, 3-amino-4-fluoro, tos-bb-0112, wfspevfsrutrcn-uhfffaoysa
PubChem CID 75396
IUPAC Name 3-amino-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)N)F

Specifications

Melting Point 182°C to 184°C
Color White
Quantity 5 g
Formula Weight 155.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Amino-4-fluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'