missing translation for 'onlineSavingsMsg'
Learn More

3-amino-4-hydroxy-5-nitrobenzene-1-sulfonic acid hydrate, Thermo Scientific™

£18.40 - £55.20

Chemical Identifiers

CAS 175278-60-1
Molecular Formula C6H8N2O7S
Molecular Weight (g/mol) 252.197
InChI Key HJASIPICABEOAV-UHFFFAOYSA-N
Synonym 3-amino-4-hydroxy-5-nitrobenzenesulfonic acid hydrate, 3-amino-4-hydroxy-5-nitrobenzene-1-sulfonic acid hydrate, benzenesulfonic acid,3-amino-4-hydroxy-5-nitro-, hydrate 1:1, benzenesulfonic acid,3-amino-4-hydroxy-5-nitro-,hydrate 1:1, 3-amino-4-hydroxy-5-nitrobenzene-1-sulfonic acid-water 1/1
PubChem CID 5712259
IUPAC Name 3-amino-4-hydroxy-5-nitrobenzenesulfonic acid;hydrate
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)N)S(=O)(=O)O.O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10566015
SDS View Documents
Thermo Scientific Maybridge
SB01747DE
5 g
£18.40
5g
Please to purchase this item. Need a web account? Register with us today!
10775473
SDS View Documents
Thermo Scientific Maybridge
SB01747EA
10 g
£31.10
10g
Please to purchase this item. Need a web account? Register with us today!
11356935
SDS View Documents
Thermo Scientific Maybridge
SB01747EB
25 g
£55.20
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-amino-4-hydroxy-5-nitrobenzene-1-sulfonic acid hydrate, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.