missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-hydroxybenzonitrile, 97%, Thermo Scientific Chemicals

Product Code. 15406737
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14543-43-2
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00234273
InChI Key ZEWCASRNRWXXSO-UHFFFAOYSA-N
PubChem CID 271258
IUPAC Name 3-amino-4-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1C#N)N)O

Specifications

Melting Point 151°C to 153°C
Quantity 250 mg
Sensitivity Air Sensitive
Synonym 2-amino-4-cyanophenol, 2-amino-4-cyano-phenol, benzonitrile, 3-amino-4-hydroxy, 3-amino-4-hydroxybenzenecarbonitrile, 5-cyano-2-hydroxyaniline, acmc-209cv1, 3-amino-4-hydroxy-benzonitrile, benzonitrile,3-amino-4-hydroxy, 5-cyano-2-hydroxy-aniline
Formula Weight 134.14
Percent Purity 97%
Chemical Name or Material 3-Amino-4-hydroxybenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'