missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-iodo-1H-indazole, 95%, Thermo Scientific™

Product Code. 10311730
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 599191-73-8
Molecular Formula C7H6IN3
Molecular Weight (g/mol) 259.05
MDL Number MFCD03426697
InChI Key OEQIVIYSUJXCFG-UHFFFAOYSA-N
Synonym 4-iodo-1h-indazol-3-ylamine, 3-amino-4-iodo-1h-indazole, 1h-indazol-3-amine, 4-iodo, 3-amino-4-iodoindazole, 4-iodo-1h-indazol-amine, ksc268o0n, 4-iodo-1h-indazole-3-amine, 4-iodo-1h-indazole-3-ylamine, 3-amino-4-iodo-indazole
PubChem CID 4057787
IUPAC Name 4-iodo-1H-indazol-3-amine
SMILES NC1=NNC2=CC=CC(I)=C12

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 150°C to 155°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Formula Weight 259.05
Percent Purity ≥94.0%
Physical Form Crystalline Solid
Chemical Name or Material 3-Amino-4-iodo-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-4-iodo-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'