missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylbenzamide, 97%, Thermo Scientific™

Product Code. 11417056
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19406-86-1
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00035936
InChI Key VYBKAZXQKUFAHG-UHFFFAOYSA-N
Synonym benzamide, 3-amino-4-methyl, 3-amino-p-toluamide, 3-amino-4-methyl-benzamide, unii-51vb4677ff, atad, acmc-1c8qe, dsstox_cid_24407, dsstox_rid_80206, 3-amino-4-methyl benzamide, 3-azanyl-4-methyl-benzamide
PubChem CID 88043
IUPAC Name 3-amino-4-methylbenzamide
SMILES CC1=C(C=C(C=C1)C(=O)N)N

Specifications

Melting Point 127°C to 131°C
Quantity 25 g
Sensitivity Air Sensitive
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 3-Amino-4-methylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'