Learn More
3-Amino-6-morpholinopyridazine, 97%, Thermo Scientific Chemicals
£136.00 - £384.00
Chemical Identifiers
CAS | 66346-91-6 |
---|---|
Molecular Formula | C8H12N4O |
Molecular Weight (g/mol) | 180.21 |
InChI Key | IEUHTZVAUDMKQJ-UHFFFAOYSA-N |
Synonym | 6-morpholinopyridazin-3-amine, 3-amino-6-morpholin-4-yl pyridazine, 6-morpholin-4-yl pyridazin-3-amine, 3-amino-6-morpholin-4-ylpyridazine, 6-morpholin-4-yl-pyridazin-3-ylamine, 3-pyridazinamine, 6-4-morpholinyl, 6-morpholinopyridazine-3-amine, 6-4-morpholinyl-3-pyridazinamine, 3-amino-6-morpholin-4-3-amino-6-morpholin-4-ylpyridazine |
PubChem CID | 13114295 |
IUPAC Name | 6-morpholin-4-ylpyridazin-3-amine |
SMILES | C1COCCN1C2=NN=C(C=C2)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11996641
|
Thermo Scientific Acros
433270010 |
1 g |
£136.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11916651
|
Thermo Scientific Acros
433270050 |
5 g |
£384.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
66346-91-6 | |
180.21 | |
6-morpholinopyridazin-3-amine, 3-amino-6-morpholin-4-yl pyridazine, 6-morpholin-4-yl pyridazin-3-amine, 3-amino-6-morpholin-4-ylpyridazine, 6-morpholin-4-yl-pyridazin-3-ylamine, 3-pyridazinamine, 6-4-morpholinyl, 6-morpholinopyridazine-3-amine, 6-4-morpholinyl-3-pyridazinamine, 3-amino-6-morpholin-4-3-amino-6-morpholin-4-ylpyridazine | |
6-morpholin-4-ylpyridazin-3-amine |
C8H12N4O | |
IEUHTZVAUDMKQJ-UHFFFAOYSA-N | |
13114295 | |
C1COCCN1C2=NN=C(C=C2)N |
Specifications
66346-91-6 | |
100.0 | |
White to Yellow | |
96% min. (HPLC) | |
C8H12N4O | |
IEUHTZVAUDMKQJ-UHFFFAOYSA-N | |
6-morpholin-4-ylpyridazin-3-amine | |
13114295 | |
97% | |
3-Amino-6-morpholinopyridazine |
96.0 | |
181°C to 183°C | |
Authentic | |
Glass Bottle | |
6-morpholinopyridazin-3-amine, 3-amino-6-morpholin-4-yl pyridazine, 6-morpholin-4-yl pyridazin-3-amine, 3-amino-6-morpholin-4-ylpyridazine, 6-morpholin-4-yl-pyridazin-3-ylamine, 3-pyridazinamine, 6-4-morpholinyl, 6-morpholinopyridazine-3-amine, 6-4-morpholinyl-3-pyridazinamine, 3-amino-6-morpholin-4-3-amino-6-morpholin-4-ylpyridazine | |
C1COCCN1C2=NN=C(C=C2)N | |
180.21 | |
180.21 | |
Crystalline Powder or Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
- 3-Amino-6-morpholinopyridazine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only