missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Aminoacetanilide, 97%, Thermo Scientific™
Click to view available options
Quantity:
50 g
250 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3′-Aminoacetanilide is mainly used as reaction intermediates dyes and disperse dyes.
Solubility
Soluble in water 1-5 g/100 mL at 24°C.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
3′-Aminoacetanilide is mainly used as reaction intermediates dyes and disperse dyes.
Solubility
Soluble in water 1-5 g/100 mL at 24°C.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
Melting Point | 86°C to 88°C |
Quantity | 50 g |
Solubility Information | Soluble in water 1-5g/100 mL at 24°C. |
Formula Weight | 150.18 |
Percent Purity | 97% |
Chemical Name or Material | 3'-Aminoacetanilide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'