Learn More
3'-Aminoacetanilide, 97%, Thermo Scientific™
£23.71 - £41.90
Chemical Identifiers
CAS | 102-28-3 |
---|---|
Molecular Formula | C8H10N2O |
Molecular Weight (g/mol) | 150.18 |
MDL Number | MFCD00025229 |
InChI Key | PEMGGJDINLGTON-UHFFFAOYSA-N |
Synonym | n-3-aminophenyl acetamide, 3'-aminoacetanilide, 3-aminoacetanilide, m-aminoacetanilide, acetamide, n-3-aminophenyl, 3-acetamidoaniline, m-acetaminoaniline, m-acetylamino aniline, 3-aminoacetanilid, 3-amino-n-acetylaniline |
PubChem CID | 7604 |
IUPAC Name | N-(3-aminophenyl)acetamide |
SMILES | CC(=O)NC1=CC=CC(N)=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3′-Aminoacetanilide is mainly used as reaction intermediates dyes and disperse dyes.
Solubility
Soluble in water 1-5 g/100 mL at 24°C.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
102-28-3 | |
150.18 | |
PEMGGJDINLGTON-UHFFFAOYSA-N | |
7604 | |
CC(=O)NC1=CC=CC(N)=C1 |
C8H10N2O | |
MFCD00025229 | |
n-3-aminophenyl acetamide, 3'-aminoacetanilide, 3-aminoacetanilide, m-aminoacetanilide, acetamide, n-3-aminophenyl, 3-acetamidoaniline, m-acetaminoaniline, m-acetylamino aniline, 3-aminoacetanilid, 3-amino-n-acetylaniline | |
N-(3-aminophenyl)acetamide |
Specifications
102-28-3 | |
C8H10N2O | |
n-3-aminophenyl acetamide, 3'-aminoacetanilide, 3-aminoacetanilide, m-aminoacetanilide, acetamide, n-3-aminophenyl, 3-acetamidoaniline, m-acetaminoaniline, m-acetylamino aniline, 3-aminoacetanilid, 3-amino-n-acetylaniline | |
PEMGGJDINLGTON-UHFFFAOYSA-N | |
N-(3-aminophenyl)acetamide | |
7604 | |
97% |
86°C to 88°C | |
MFCD00025229 | |
Soluble in water 1-5g/100 mL at 24°C. | |
CC(=O)NC1=CC=CC(N)=C1 | |
150.18 | |
150.18 | |
3'-Aminoacetanilide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 203-021-5
missing translation for 'rtecsNumber' : AD8050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures