missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoazetidine dihydrochloride, 95%

Product Code. 15495588
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
This item is not returnable. View return policy

Product Code. 15495588

Brand: Thermo Scientific Alfa Aesar H59318.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102065-89-4 | C3H8N2 | 72.111 g/mol

3-Aminoazetidine dihydrochloride, is used as active pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminoazetidine dihydrochloride, is used as active pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102065-89-4
Molecular Formula C3H8N2
Molecular Weight (g/mol) 72.111
MDL Number MFCD09910173
InChI Key FDPKMJDUXJFKOI-UHFFFAOYSA-N
Synonym 3-aminoazetidine, 3-azetidinamine, acmc-209y1n, 3-aminoazetidine dihy drochloric
PubChem CID 1516506
IUPAC Name azetidin-3-amine
SMILES C1C(CN1)N

Specifications

Quantity 500 mg
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 145.03
Percent Purity 95%
Chemical Name or Material 3-Aminoazetidine dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.