missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzotrifluoride, 98%, Thermo Scientific Chemicals

Product Code. 10725361
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-16-8
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.13
MDL Number MFCD00007797
InChI Key VIUDTWATMPPKEL-UHFFFAOYSA-N
Synonym 3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline
PubChem CID 7375
IUPAC Name 3-(trifluoromethyl)aniline
SMILES C1=CC(=CC(=C1)N)C(F)(F)F

Specifications

CAS Min % 98
Melting Point 5.0°C to 6.0°C
Color Yellow
Density 1.3030g/mL
Boiling Point 187.0°C to 193.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CF3
Refractive Index 1.4790 to 1.4810
Quantity 5 mL
Beilstein 12, 870
Specific Gravity 1.303
Solubility Information Solubility in water: 5g/L in water (20°C). Other solubilities: soluble in ethanol,ether and diethyl ether
Formula Weight 161.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Aminobenzotrifluoride, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Aminobenzotrifluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.