Learn More
3-Aminobenzyl alcohol, 97%, Thermo Scientific Chemicals
£37.40 - £112.00
Chemical Identifiers
CAS | 1877-77-6 |
---|---|
Molecular Formula | C7H9NO |
Molecular Weight (g/mol) | 123.16 |
MDL Number | MFCD00007817 |
InChI Key | OJZQOQNSUZLSMV-UHFFFAOYSA-N |
Synonym | 3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol |
PubChem CID | 80293 |
IUPAC Name | (3-aminophenyl)methanol |
SMILES | NC1=CC=CC(CO)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10387600
|
Thermo Scientific Acros
184570050 |
5 g | Glass bottle |
£37.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10390254
|
Thermo Scientific Acros
184570250 |
25 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1877-77-6 | |
123.16 | |
OJZQOQNSUZLSMV-UHFFFAOYSA-N | |
80293 | |
NC1=CC=CC(CO)=C1 |
C7H9NO | |
MFCD00007817 | |
3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol | |
(3-aminophenyl)methanol |
Specifications
1877-77-6 | |
100.0 | |
Beige or Beige-Gray to Brown | |
96% min. (GC) | |
H2NC6H4CH2OH | |
13, 619 | |
OJZQOQNSUZLSMV-UHFFFAOYSA-N | |
(3-aminophenyl)methanol | |
80293 | |
97% | |
3-Aminobenzyl alcohol |
96.0 | |
91.0°C to 95.0°C | |
Authentic | |
C7H9NO | |
MFCD00007817 | |
3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol | |
NC1=CC=CC(CO)=C1 | |
123.16 | |
123.15 | |
Fine Crystalline Powder |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 217-514-8
- 3-Aminobenzyl alcohol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only