missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylboronic acid hydrochloride, 98%, Thermo Scientific Chemicals

£64.40 - £193.00

Chemical Identifiers

CAS 85006-23-1
Molecular Formula C6H9BClNO2
Molecular Weight (g/mol) 173.40
MDL Number MFCD00191748
InChI Key QBMHZZHJIBUPOX-UHFFFAOYSA-N
Synonym 3-aminophenyl boronic acid hydrochloride, 3-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid hcl, m-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid, hcl, 3-aminobenzeneboronic acid hydrochloride, boronic acid, 3-aminophenyl-, hydrochloride, pubchem1751, pubchem20396, timtec-bb sbb003816
PubChem CID 2763241
IUPAC Name (3-aminophenyl)boronic acid;hydrochloride
SMILES Cl.NC1=CC=CC(=C1)B(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10537410
View Documents
Thermo Scientific Acros
435530010
1 g
£64.40
1g
Please to purchase this item. Need a web account? Register with us today!
10027909
View Documents
Thermo Scientific Acros
435530050
5 g
£193.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

85006-23-1
173.40
QBMHZZHJIBUPOX-UHFFFAOYSA-N
2763241
Cl.NC1=CC=CC(=C1)B(O)O
C6H9BClNO2
MFCD00191748
3-aminophenyl boronic acid hydrochloride, 3-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid hcl, m-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid, hcl, 3-aminobenzeneboronic acid hydrochloride, boronic acid, 3-aminophenyl-, hydrochloride, pubchem1751, pubchem20396, timtec-bb sbb003816
(3-aminophenyl)boronic acid;hydrochloride

Specifications

85006-23-1
100.0
40.52 to 42.60% C, 7.88 to 8.28% N
98% min. (HPLC)
C6H9BClNO2
3-aminophenyl boronic acid hydrochloride, 3-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid hcl, m-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid, hcl, 3-aminobenzeneboronic acid hydrochloride, boronic acid, 3-aminophenyl-, hydrochloride, pubchem1751, pubchem20396, timtec-bb sbb003816
Cl.NC1=CC=CC(=C1)B(O)O
173.40
173.41
Crystalline Powder
97.5
White to Gray
Authentic
Glass Bottle
MFCD00191748
QBMHZZHJIBUPOX-UHFFFAOYSA-N
(3-aminophenyl)boronic acid;hydrochloride
2763241
98%
3-Aminophenylboronic acid hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 285-052-4

Product Identifier
  • 3-Aminophenylboronic acid hydrochloride, 98%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only