missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopiperidine dihydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 10336404
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138060-07-8
Molecular Formula C5H12N2·2HCl
Molecular Weight (g/mol) 173.09
MDL Number MFCD00012773
InChI Key GGPNYXIOFZLNKW-UHFFFAOYSA-N
Synonym 3-aminopiperidine dihydrochloride, piperidin-3-amine dihydrochloride, 3-aminopiperidine 2hcl, 3-aminopiperidine hcl, 3-piperidinamine dihydrochloride, 3-piperidinamine, dihydrochloride, pubchem6755, acmc-209tvl, ksc839o8j, 3-amino-piperidine di-hydrochloride
PubChem CID 16211870
IUPAC Name piperidin-3-amine;dihydrochloride
SMILES C1CC(CNC1)N.Cl.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 230.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 173.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Aminopiperidine dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminopiperidine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'