missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopropionitrile, 98%, stab. with potassium carbonate, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.
Solubility
Soluble in water.
Notes
It is air and moisture sensitive. Incompatible with oxidizing agents, acids and heat.
3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.
Solubility
Soluble in water.
Notes
It is air and moisture sensitive. Incompatible with oxidizing agents, acids and heat.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.952 |
Boiling Point | 79°C to 81°C (16 mmHg) |
Odor | Amine-like |
Linear Formula | H2NCH2CH2CN |
Refractive Index | 1.4375 |
Quantity | 25 g |
UN Number | UN2735 |
Beilstein | 1698848 |
Sensitivity | Air and moisture sensitive |
Merck Index | 14,469 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'