missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopyridine, 99%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
250 g
1000 g
Description
3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.
Solubility
It is soluble in water as well as soluble in alcohol, benzene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under ordinary conditions.
3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.
Solubility
It is soluble in water as well as soluble in alcohol, benzene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under ordinary conditions.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 58°C to 63°C |
pH | 10 |
Boiling Point | 250°C to 252°C |
Flash Point | 88°C (190°F) |
Odor | Unpleasant |
Quantity | 50 g |
UN Number | UN2671 |
Beilstein | 105692 |
Sensitivity | Hygroscopic |
Merck Index | 14,473 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'