missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoquinoline, 99%, Thermo Scientific Chemicals

Product Code. 10616914
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 580-17-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00006772
InChI Key SVNCRRZKBNSMIV-UHFFFAOYSA-N
Synonym 3-aminoquinoline, 3-quinolinamine, 3-quinolylamine, 3-quinolineamine, quinoline, 3-amino, quinolin-3-ylamine, 3-amino quinoline, unii-98ovl5k3r4, quinolin-3-yl-amine, ccris 1678
PubChem CID 11375
IUPAC Name quinolin-3-amine
SMILES C1=CC=C2C(=C1)C=C(C=N2)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 91.0°C to 94.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 22, I, 638
Solubility Information Solubility in water: slightly soluble in hot water. Other solubilities: soluble in methanol
Formula Weight 144.18
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Aminoquinoline, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'