missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoquinuclidine dihydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10173140
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 6530-09-2
Molecular Formula C7H14N2·2ClH
Molecular Weight (g/mol) 199.12
MDL Number MFCD00137395
InChI Key STZHBULOYDCZET-UHFFFAOYNA-N
ChEBI CHEBI:47953

Especificaciones

CAS Min % 97.5
CAS Max % 100.0
Melting Point 321°C to 323°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Synonym 2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
Solubility Information Solubility in water: soluble
SMILES [H+].[H+].[Cl-].[Cl-].NC1CN2CCC1CC2
IUPAC Name (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
PubChem CID 65041
Formula Weight 199.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Aminoquinuclidine dihydrochloride, 98%
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 3-Aminoquinuclidine dihydrochloride
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'