missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Azido-1-propylamine, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
1 g
Description
Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.
Solubility
Soluble in DMSO, DMF, DCM, chloroform, THF, and water.Miscible with dimethylsulfoxide, dimethyl formamide, dichloromethane, tetrahydrofuran, chloroform and water.
Notes
Incompatible with strong oxidizing agents.
Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.
Solubility
Soluble in DMSO, DMF, DCM, chloroform, THF, and water.Miscible with dimethylsulfoxide, dimethyl formamide, dichloromethane, tetrahydrofuran, chloroform and water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Color | Yellow |
Flash Point | 60°C (140°F) |
Linear Formula | N3CH2CH2CH2NH2 |
Quantity | 100 mg |
UN Number | UN1993 |
Solubility Information | Soluble in DMSO,DMF,DCM,chloroform,THF,and water;Miscible with dimethylsulfoxide,dimethyl formamide,dichloromethane,tetrahydrofuran,chloroform and water. |
Formula Weight | 100.12 |
Physical Form | Liquid |
Chemical Name or Material | 3-Azido-1-propylamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'