missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 3'-Azido-3'-deoxythymidine, 98%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3'-Azido-3'-deoxythymidine is a Anti-viral agent, and also used as a reverse transcriptase inhibitor active against HIV-1 virus that blocks the incorporation of nucleotides into growing DNA strands. Useful for antiviral and anticancer studies. 3?-Azido-3?-deoxythymidine enters CNS by diffusion and causes irreversible telomere shortening.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3′-Azido-3′-deoxythymidine is a Anti-viral agent, and also used as a reverse transcriptase inhibitor active against HIV-1 virus that blocks the incorporation of nucleotides into growing DNA strands. Useful for antiviral and anticancer studies. 3′-Azido-3′-deoxythymidine enters CNS by diffusion and causes irreversible telomere shortening.
Solubility
Soluble in water.
Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
3′-Azido-3′-deoxythymidine is a Anti-viral agent, and also used as a reverse transcriptase inhibitor active against HIV-1 virus that blocks the incorporation of nucleotides into growing DNA strands. Useful for antiviral and anticancer studies. 3′-Azido-3′-deoxythymidine enters CNS by diffusion and causes irreversible telomere shortening.
Solubility
Soluble in water.
Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | 113°C to 115°C |
Color | White |
Quantity | 1 g |
Beilstein | 3595791 |
Sensitivity | Light sensitive; Very Hygroscopic |
Merck Index | 1410123 |
Solubility Information | Soluble in water. |
IUPAC Name | 1-[(2R,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione |
Formula Weight | 267.24 |
Percent Purity | 98% |
Show More |
Safety and Handling
Signal Word
- Warning
- Carcinogenicity Category 2
- H351-Suspected of causing cancer.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'