missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Benzoylbenzoic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
3-Benzoylbenzoic acid has been used in light-induced reactions on benzophenone-terminated boron-doped diamond (BDD) surfaces and preparation of benzophenone flavonol derivative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Benzoylbenzoic acid has been used in light-induced reactions on benzophenone-terminated boron-doped diamond (BDD) surfaces and preparation of benzophenone flavonol derivative.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
3-Benzoylbenzoic acid has been used in light-induced reactions on benzophenone-terminated boron-doped diamond (BDD) surfaces and preparation of benzophenone flavonol derivative.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 162°C to 166°C |
Odor | Odorless |
Quantity | 5 g |
Beilstein | 2212366 |
Synonym | benzoic acid, 3-benzoyl, m-benzoylbenzoic acid, 3-benzoyl benzoic acid, 3-benzoyl-benzoic acid, 3-phenylcarbonyl benzoic acid, 3-benzoylbenzoicacid, 3-benzoyl-benzoesaure, 3-benzoylbenzoic-acid, acmc-20an9k, benzoic acid,3-benzoyl |
Solubility Information | Insoluble in water. |
Formula Weight | 226.23 |
Percent Purity | 98% |
Chemical Name or Material | 3-Benzoylbenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'