missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxy-4-methoxybenzoic acid, 97+%, Thermo Scientific™

Product Code. 11317585
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Benzyloxy-4-methoxybenzoic acid is used as Pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58452-00-9
Molecular Formula C15H14O4
Molecular Weight (g/mol) 258.27
MDL Number MFCD00016526
InChI Key YPDXIGBSOBESNI-UHFFFAOYSA-N
PubChem CID 94057
SMILES COC1=CC=C(C=C1OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 177°C to 182°C
Quantity 5 g
Beilstein 2660886
Synonym 3-benzyloxy-4-methoxybenzoic acid, 3-benzyloxy-p-anisic acid, 3-benzyloxy-4-methoxy benzoic acid, 4-methoxy-3-phenylmethoxy benzoic acid, benzoic acid, 4-methoxy-3-phenylmethoxy, acmc-20an9h, 3-benzyloxy-4-methoxy-benzoic acid, benzoic acid, 3-benzyloxy-4-methoxy, benzoic acid,4-methoxy-3-phenylmethoxy
Solubility Information Insoluble in water.
IUPAC Name 4-methoxy-3-phenylmethoxybenzoic acid
Formula Weight 258.28
Percent Purity ≥97%
Chemical Name or Material 3-Benzyloxy-4-methoxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'