missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Benzyloxybenzaldehyde, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 55°C to 59°C |
Color | White to Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C14H12O2 |
Linear Formula | C6H5CH2OC6H4CHO |
Quantity | 5 g |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'