missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzoyl chloride, 95%, Thermo Scientific Chemicals

£25.90 - £257.28

Chemical Identifiers

CAS 61535-46-4
Molecular Formula C14H11ClO2
Molecular Weight (g/mol) 246.69
MDL Number MFCD02258040
InChI Key HPVUGOBQRQUWMK-UHFFFAOYSA-N
Synonym 3-benzyloxy benzoyl chloride, 3-benzyloxybenzoyl chloride, 3-benzyloxy-benzoyl chloride, 3-benzyl-oxy benzoyl chloride, 3-phenylmethoxy benzoyl chloride, acmc-20ah7b, 3-benzyloxyl-benzoyl chloride, 3-benzyloxy-benzoic acid choride, benzoyl chloride, 3-phenylmethoxy
PubChem CID 4995375
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11355419
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50410.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11365419
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50410.03
1 g
£83.67
1g
Please to purchase this item. Need a web account? Register with us today!
15407877
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50410.06
5 g
£257.28
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reactant for preparation of dihydroquinolinones and tetrahydroquinazolinones as potent kinesin spindle protein inhibitors and potential antitumor agents, preparation of carbamate derivatives as fatty acid amide hydrolase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for preparation of dihydroquinolinones and tetrahydroquinazolinones as potent kinesin spindle protein inhibitors and potential antitumor agents, preparation of carbamate derivatives as fatty acid amide hydrolase inhibitors.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

61535-46-4
246.69
HPVUGOBQRQUWMK-UHFFFAOYSA-N
4995375
C14H11ClO2
MFCD02258040
3-benzyloxy benzoyl chloride, 3-benzyloxybenzoyl chloride, 3-benzyloxy-benzoyl chloride, 3-benzyl-oxy benzoyl chloride, 3-phenylmethoxy benzoyl chloride, acmc-20ah7b, 3-benzyloxyl-benzoyl chloride, 3-benzyloxy-benzoic acid choride, benzoyl chloride, 3-phenylmethoxy

Specifications

61535-46-4
C14H11ClO2
UN3261
3-benzyloxy benzoyl chloride, 3-benzyloxybenzoyl chloride, 3-benzyloxy-benzoyl chloride, 3-benzyl-oxy benzoyl chloride, 3-phenylmethoxy benzoyl chloride, acmc-20ah7b, 3-benzyloxyl-benzoyl chloride, 3-benzyloxy-benzoic acid choride, benzoyl chloride, 3-phenylmethoxy
HPVUGOBQRQUWMK-UHFFFAOYSA-N
3-phenylmethoxybenzoyl chloride
4995375
95%
40°C to 42°C
MFCD02258040
Moisture sensitive
Reacts with water.
C1=CC=C(C=C1)COC2=CC=CC(=C2)C(=O)Cl
246.69
246.7
3-Benzyloxybenzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only