missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Benzyloxyphenylacetic acid, 95%, Thermo Scientific™
£43.90 - £138.00
Chemical Identifiers
CAS | 1860-58-8 |
---|---|
Molecular Formula | C15H14O3 |
Molecular Weight (g/mol) | 242.27 |
MDL Number | MFCD02664806 |
InChI Key | LLZKAZNUCYYBQO-UHFFFAOYSA-N |
Synonym | 2-3-benzyloxy phenyl acetic acid, 3-benzyloxyphenylacetic acid, 3-benzyloxy-phenyl-acetic acid, 3-benzyloxyphenyl acetic acid, 3-benzyloxy phenyl acetic acid, 2-3-phenylmethoxyphenyl acetic acid, 2-3-phenylmethoxy phenyl acetic acid, acmc-1bpe0, 3-benzyloxybenzeneacetic acid |
PubChem CID | 4592030 |
IUPAC Name | 2-(3-phenylmethoxyphenyl)acetic acid |
SMILES | OC(=O)CC1=CC(OCC2=CC=CC=C2)=CC=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1860-58-8 | |
242.27 | |
LLZKAZNUCYYBQO-UHFFFAOYSA-N | |
4592030 | |
OC(=O)CC1=CC(OCC2=CC=CC=C2)=CC=C1 |
C15H14O3 | |
MFCD02664806 | |
2-3-benzyloxy phenyl acetic acid, 3-benzyloxyphenylacetic acid, 3-benzyloxy-phenyl-acetic acid, 3-benzyloxyphenyl acetic acid, 3-benzyloxy phenyl acetic acid, 2-3-phenylmethoxyphenyl acetic acid, 2-3-phenylmethoxy phenyl acetic acid, acmc-1bpe0, 3-benzyloxybenzeneacetic acid | |
2-(3-phenylmethoxyphenyl)acetic acid |
Specifications
1860-58-8 | |
MFCD02664806 | |
LLZKAZNUCYYBQO-UHFFFAOYSA-N | |
2-(3-phenylmethoxyphenyl)acetic acid | |
4592030 | |
95% |
C15H14O3 | |
2-3-benzyloxy phenyl acetic acid, 3-benzyloxyphenylacetic acid, 3-benzyloxy-phenyl-acetic acid, 3-benzyloxyphenyl acetic acid, 3-benzyloxy phenyl acetic acid, 2-3-phenylmethoxyphenyl acetic acid, 2-3-phenylmethoxy phenyl acetic acid, acmc-1bpe0, 3-benzyloxybenzeneacetic acid | |
OC(=O)CC1=CC(OCC2=CC=CC=C2)=CC=C1 | |
242.27 | |
242.28 | |
3-Benzyloxyphenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures