missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)-3-phenylpropionic acid, 97%

Product Code. 15409109
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15409109

Brand: Thermo Scientific Alfa Aesar L19802.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14676-01-8 | C14H19NO4 | 265.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 14676-01-8
Melting Point 142°C to 144°C (decomposition)
Molecular Formula C14H19NO4
MDL Number MFCD02090695
Quantity 1 g
Beilstein 2867023
Synonym 3-boc-amino-3-phenylpropionic acid, 3-n-boc-3-phenylpropionic acid, boc-dl-beta-phe-oh, 3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, 3-boc-amino-3-phenyl-propionic acid, 3-tert-butoxycarbonylamino-3-phenylpropanoic acid, 3-tert-butoxy carbonyl amino-3-phenylpropanoic acid, boc-d-, a-phe-oh, pubchem16181
InChI Key JTNQFJPZRTURSI-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1
IUPAC Name 3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
Molecular Weight (g/mol) 265.309
PubChem CID 2756815
Formula Weight 265.31
Percent Purity 97%
Chemical Name or Material 3-(Boc-amino)-3-phenylpropionic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.