missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)phenol, 97%, Thermo Scientific Chemicals

Product Code. 15401188
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19962-06-2
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.25
MDL Number MFCD06798070
InChI Key HJQNVUQTARSZDK-UHFFFAOYSA-N
Synonym tert-butyl 3-hydroxyphenylcarbamate, 3-boc-amino phenol, tert-butyl n-3-hydroxyphenyl carbamate, tert-butyl 3-hydroxyphenyl carbamate, n-boc-3-aminophenol, carbamic acid, 3-hydroxyphenyl-, 1,1-dimethylethyl ester, carbamic acid, n-3-hydroxyphenyl-, 1,1-dimethylethyl ester, carbamic acid,n-3-hydroxyphenyl-, 1,1-dimethylethyl ester, tert-butoxy-n-3-hydroxyphenyl carboxamide, n-boc-3-amino-phenol
PubChem CID 312485
IUPAC Name tert-butyl N-(3-hydroxyphenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=CC(O)=C1

Specifications

Melting Point 134°C to 138°C
Quantity 1 g
Formula Weight 209.25
Percent Purity 97%
Chemical Name or Material 3-(Boc-amino)phenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'