missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)piperidine, 97%, Thermo Scientific Chemicals

Product Code. p-7035083
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15419537

Brand: Thermo Scientific Alfa Aesar H52788.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172603-05-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03839941
InChI Key WUOQXNWMYLFAHT-UHFFFAOYSA-N
Synonym 3-boc-aminopiperidine, tert-butyl piperidin-3-ylcarbamate, 3-n-boc-aminopiperidine, tert-butyl n-piperidin-3-yl carbamate, 3-boc-amino piperidine, 3-n-boc-amino piperidine, 3-tert-butoxycarbonylamino piperidine, 3-aminopiperidine, 3-boc protected, piperidin-3-yl-carbamic acid tert-butyl ester, tert-butyl n-3-piperidyl carbamate
PubChem CID 4233041
IUPAC Name tert-butyl N-piperidin-3-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1CCCNC1

Specifications

Quantity 1 g
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material 3-(Boc-amino)piperidine
compliance-icons
Product Identifier
  • 3-(Boc-amino)piperidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 3-(BOC-Amino)piperidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.