missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(BOC-Aminomethyl)benzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Melting Point | 129.0°C to 134.8°C |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | White |
Assay Percent Range | 96% min. (HPLC) |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Formula Weight | 251.28 |
Percent Purity | 97% |
Quantity | 5 g |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'