missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1,2-propanediol, 97%, Thermo Scientific Chemicals

Product Code. 10562801
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4704-77-2
Molecular Formula C3H7BrO2
Molecular Weight (g/mol) 154.99
MDL Number MFCD00004711
InChI Key SIBFQOUHOCRXDL-UHFFFAOYSA-N
PubChem CID 20827
ChEBI CHEBI:18719
IUPAC Name 3-bromopropane-1,2-diol
SMILES C(C(CBr)O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.7700g/mL
Boiling Point 72.0°C to 75.0°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CH(OH)CH2OH
Refractive Index 1.5165 to 1.5195
Quantity 25 mL
Beilstein 01, I, 248
Specific Gravity 1.77
Synonym 3-bromo-1,2-propanediol, monobromoglycerol, alpha-bromohydrin, 1,2-propanediol, 3-bromo, bromodeoxyglycerol, glycerol bromohydrin, 1-bromo-2,3-propanediol, 3-bromodeoxyglycerol, rs-3-bromo-1,2-propanediol, 1,2-propanediol,3-bromo
Formula Weight 154.99
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material 3-Bromo-1, 2-propanediol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-1,2-propanediol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'