missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-propanol, 92%, Thermo Scientific Chemicals

Product Code. 10463073
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-18-9
Molecular Formula C3H7BrO
Molecular Weight (g/mol) 138.99
MDL Number MFCD00002942
InChI Key RQFUZUMFPRMVDX-UHFFFAOYSA-N
Synonym 3-bromo-1-propanol, trimethylene bromohydrin, 1-propanol, 3-bromo, 3-bromopropanol, 3-hydroxypropyl bromide, 1-bromo-3-propanol, 3-bromo-propan-1-ol, ccris 5979, 1-bromo-3-hydroxypropane, unii-3t4axh68fh
PubChem CID 12308
IUPAC Name 3-bromopropan-1-ol
SMILES OCCCBr

Specifications

CAS Min % 90.0
CAS Max % 100.0
Density 1.6000g/mL
Boiling Point 62.0°C (5.0 mmHg)
Flash Point 65°C
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)3OH
Refractive Index 1.4860 to 1.4900
Quantity 5 g
Beilstein 01, 356
Specific Gravity 1.6
Solubility Information Solubility in water: 167g/L water (20°C). Other solubilities: slightly soluble in ethanol and ether, soluble in most solvents
Formula Weight 138.99
Percent Purity 92%
Chemical Name or Material 3-Bromo-1-propanol, 92%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-1-propanol, 92%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.