missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.
Solubility
Soluble in dimethylsulphoxide and ethanol. Insoluble in water.
Notes
Incompatible with oxidizing agents.
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.
Solubility
Soluble in dimethylsulphoxide and ethanol. Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Melting Point | 192°C to 196°C |
Quantity | 1 g |
Solubility Information | Soluble in dimethylsulphoxide and ethanol. Insoluble in water. |
Formula Weight | 387.24 |
Percent Purity | ≥98% |
Chemical Name or Material | (3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'