missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 103192-90-1 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C6H2BrF4N |
Quantity | 1 g |
Formula Weight | 243.98 |
Percent Purity | 97% |
Chemical Name or Material | 3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'