missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-furoic acid, 97%, Thermo Scientific™

Product Code. 10441884
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 14903-90-3
Molecular Formula C5H3BrO3
Molecular Weight (g/mol) 190.98
MDL Number MFCD00092761
InChI Key UZBGSJZFBUOJNE-UHFFFAOYSA-N
PubChem CID 605479
IUPAC Name 3-bromofuran-2-carboxylic acid
SMILES C1=COC(=C1Br)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-bromo-2-furoic acid, furancarboxylic acid, bromo, 3-bromo-2-furancarboxylic acid, 3-bromo-furan-2-carboxylic acid, 3-bromofuran-2-carboxylicacid, bromofuroic acid, 3-bomo-2-furoic acid, 2-furancarboxylic acid,3-bromo, 3-bromanylfuran-2-carboxylic acid, 2-furancarboxylic acid, 3-bromo
Formula Weight 190.98
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-furoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'