missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-furoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 3-bromo-2-furoic acid, furancarboxylic acid, bromo, 3-bromo-2-furancarboxylic acid, 3-bromo-furan-2-carboxylic acid, 3-bromofuran-2-carboxylicacid, bromofuroic acid, 3-bomo-2-furoic acid, 2-furancarboxylic acid,3-bromo, 3-bromanylfuran-2-carboxylic acid, 2-furancarboxylic acid, 3-bromo |
Formula Weight | 190.98 |
Percent Purity | 97% |
Chemical Name or Material | 3-Bromo-2-furoic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-Bromo-2-furoic acid, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction