Learn More
3-Bromo-2-furoyl chloride, 97%, Thermo Scientific™
£98.90 - £401.00
Chemical Identifiers
CAS | 58777-57-4 |
---|---|
Molecular Formula | C5H2BrClO2 |
Molecular Weight (g/mol) | 209.423 |
MDL Number | MFCD11109330 |
InChI Key | WTZDPJVRZQGTET-UHFFFAOYSA-N |
Synonym | 3-bromo-2-furoyl chloride, 3-bromo-2-furancarbonyl chloride, 3-bromo-furan-2-carbonyl chloride, 2-furancarbonyl chloride, 3-bromo-9ci |
PubChem CID | 21864576 |
IUPAC Name | 3-bromofuran-2-carbonyl chloride |
SMILES | C1=COC(=C1Br)C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10163464
|
Thermo Scientific Maybridge
CC67202DA |
1 g | Amber Glass Bottle |
£98.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10739025
|
Thermo Scientific Maybridge
CC67202DE |
5 g | Amber Glass Bottle |
£401.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
58777-57-4 | |
209.423 | |
WTZDPJVRZQGTET-UHFFFAOYSA-N | |
21864576 | |
C1=COC(=C1Br)C(=O)Cl |
C5H2BrClO2 | |
MFCD11109330 | |
3-bromo-2-furoyl chloride, 3-bromo-2-furancarbonyl chloride, 3-bromo-furan-2-carbonyl chloride, 2-furancarbonyl chloride, 3-bromo-9ci | |
3-bromofuran-2-carbonyl chloride |
Specifications
58777-57-4 | |
100.0 | |
C5H2BrClO2 | |
3-bromo-2-furoyl chloride, 3-bromo-2-furancarbonyl chloride, 3-bromo-furan-2-carbonyl chloride, 2-furancarbonyl chloride, 3-bromo-9ci | |
C1=COC(=C1Br)C(=O)Cl | |
209.423 | |
209.43 | |
3-Bromo-2-furoyl chloride |
96.5 | |
97% | |
MFCD11109330 | |
WTZDPJVRZQGTET-UHFFFAOYSA-N | |
3-bromofuran-2-carbonyl chloride | |
21864576 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger