missing translation for 'onlineSavingsMsg'
Learn More

3-bromo-2-hydroxy-5-nitrobenzaldehyde, Thermo Scientific™

Product Code. 10507883
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10507883

Brand: Thermo Scientific Maybridge BTB03755DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16789-84-7
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.02
MDL Number MFCD00051833
InChI Key BESBCGANGAEHPM-UHFFFAOYSA-N
Synonym 3-bromo-2-hydroxy-5-nitrobenzaldehyde, 3-bromo-5-nitrosalicylaldehyde, 3-bromo-2-hydroxy-5-nitrobenzenecarbaldehyde, 2-hydroxy-3-bromo-5-nitrobenzaldehyde, 3-bromo-2-hydroxy-5-nitro-benzaldehyde, acmc-1cfaz, 3-bromo-5-nitro-salicylaldehyde, benzaldehyde,3-bromo-2-hydroxy-5-nitro, 3-bromanyl-5-nitro-2-oxidanyl-benzaldehyde
PubChem CID 519307
IUPAC Name 3-bromo-2-hydroxy-5-nitrobenzaldehyde
SMILES OC1=C(Br)C=C(C=C1C=O)[N+]([O-])=O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 246.02
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-hydroxy-5-nitrobenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.