missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4,5-dimethoxybenzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Melting Point | 61°C to 63°C |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Quantity | 1 g |
Synonym | 5-bromoveratraldehyde, 3-bromo-4,5-dimethoxy benzaldehyde, 3,4-dimethoxy-5-bromobenzaldehyde, benzaldehyde,3-bromo-4,5-dimethoxy, benzaldehyde, 3-bromo-4,5-dimethoxy, 5-bromoveratric aldehyde, acmc-209ml7, cambridge id 7187634, 5-bromveratrumaldehyde |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 245.07 |
Percent Purity | 97% |
Chemical Name or Material | 3-Bromo-4, 5-dimethoxybenzaldehyde |
Safety and Handling
Product Identifier
- 3-Bromo-4,5-dimethoxybenzaldehyde
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'